Learn More
Medchemexpress LLC Acetazolamide impurity 1 | 32873-56-6 | MFCD00128160 | 98.0% | 175.23 | C4H5N3OS2 | 1 G
SDP
Catalog No. 5000301276
Encompass_Preferred
Acetazolamide impurity 1 is an analytically characterized impurity related to acetazolamide, provided as a reference standard for method development, impurity profiling, and quality control. The product has confirmed identity (CAS 32873-56-6), molecular formula C4H5N3OS2, and molecular weight 175.23.
  • Reference standard for analytical method development and impurity profiling.
  • Suitable for quality control and stability testing.
  • Verified identity and composition for reliable traceability.
  • Supplied with a documented datasheet for characterization details.
  • Applicable to LC/GC method validation and comparative analysis.
Catalog No. 50-003-01276 Supplier Medchemexpress LLC Supplier No. HYW0286861G
Acetazolamide impurity 1 is an analytically characterized impurity related to acetazolamide, provided as a reference standard for method development, impurity profiling, and quality control. The product has confirmed identity (CAS 32873-56-6), molecular formula C4H5N3OS2, and molecular weight 175.23.
  • Reference standard for analytical method development and impurity profiling.
  • Suitable for quality control and stability testing.
  • Verified identity and composition for reliable traceability.
  • Supplied with a documented datasheet for characterization details.
  • Applicable to LC/GC method validation and comparative analysis.