Learn More
Medchemexpress LLC Acetazolamide impurity 1 | 32873-56-6 | MFCD00128160 | 98.0% | 175.23 g/mol | C4H5N3OS2 | 10 G
SDP
Catalog No. 5000301279
Encompass_Preferred
Acetazolamide impurity 1 is a characterized small-molecule impurity related to the pharmaceutical acetazolamide, provided for analytical and research purposes as an impurity reference and drug intermediate. It is intended for laboratory use only and is not for human therapeutic use.
  • Used as an analytical reference standard for impurity profiling.
  • Supports method development and quality control assays.
  • Applicable for drug intermediate and synthetic studies.
  • Stable under recommended storage; follow manufacturer storage guidance.
  • Provided as a research-grade chemical for laboratory applications.
Catalog No. 50-003-01279 Supplier Medchemexpress LLC Supplier No. HYW02868610G
Acetazolamide impurity 1 is a characterized small-molecule impurity related to the pharmaceutical acetazolamide, provided for analytical and research purposes as an impurity reference and drug intermediate. It is intended for laboratory use only and is not for human therapeutic use.
  • Used as an analytical reference standard for impurity profiling.
  • Supports method development and quality control assays.
  • Applicable for drug intermediate and synthetic studies.
  • Stable under recommended storage; follow manufacturer storage guidance.
  • Provided as a research-grade chemical for laboratory applications.