Learn More
Medchemexpress LLC Acetazolamide sodium | 1424-27-7 | 99.5% | 5 G
SDP
Catalog No. 5000409319
Encompass_Preferred
Acetazolamide sodium is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It also inhibits hCA II with an IC50 of 130 nM. This small heteroaromatic sulfonamide exhibits diuretic, antihypertensive, and anti-gonococcal activities. It is for research use only.
  • Carbonic anhydrase (CA) IX inhibitor
  • IC50 of 30 nM for hCA IX
  • Inhibits hCA II with an IC50 of 130 nM
  • Exhibits diuretic activity
  • Exhibits antihypertensive activity
  • Exhibits anti-gonococcal activity
  • Small heteroaromatic sulfonamide
Catalog No. 50-004-09319 Supplier Medchemexpress LLC Supplier No. HYB0782A5G
Acetazolamide sodium is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It also inhibits hCA II with an IC50 of 130 nM. This small heteroaromatic sulfonamide exhibits diuretic, antihypertensive, and anti-gonococcal activities. It is for research use only.
  • Carbonic anhydrase (CA) IX inhibitor
  • IC50 of 30 nM for hCA IX
  • Inhibits hCA II with an IC50 of 130 nM
  • Exhibits diuretic activity
  • Exhibits antihypertensive activity
  • Exhibits anti-gonococcal activity
  • Small heteroaromatic sulfonamide