Learn More
Medchemexpress LLC Acetazolamide (Standard) | 59-66-5 | 99.7% | 25 MG
SDP
Catalog No. 5000450807
Encompass_Preferred
Acetazolamide (Standard) is the analytical standard of Acetazolamide. It is intended for research and analytical applications. This compound acts as a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX and demonstrates diuretic, antihypertensive, and anti-gonococcal activities.
  • Analytical standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Inhibits carbonic anhydrase (CA) IX with an IC50 of 30 nM for hCA IX
  • Exhibits diuretic, antihypertensive, and anti-gonococcal activities
Catalog No. 50-004-50807 Supplier Medchemexpress LLC Supplier No. 25MGHYB0782R
Acetazolamide (Standard) is the analytical standard of Acetazolamide. It is intended for research and analytical applications. This compound acts as a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX and demonstrates diuretic, antihypertensive, and anti-gonococcal activities.
  • Analytical standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Inhibits carbonic anhydrase (CA) IX with an IC50 of 30 nM for hCA IX
  • Exhibits diuretic, antihypertensive, and anti-gonococcal activities