Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetazolamide (Standard) | 59-66-5 | 99.7% | 50 MG
SDP

Catalog No. 5000408042
Encompass_Preferred

Acetazolamide (Standard) is an analytical standard of Acetazolamide, designed for research and analytical applications. It functions as a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX and possesses diuretic, antihypertensive, and anti-gonococcal properties. This product is specifically intended for laboratory use and is not sold to patients.

  • Analytical standard for assays
  • Intended for qualitative, quantitative, and methodological research experiments
  • Commonly used in chromatography (HPLC, GC, MS)
  • High purity of 99.69%
  • Appears as a white to off-white solid
  • Molecular weight: 222.25
  • Chemical formula: C₄H₆N₄O₃S₂
  • Not for human use

Catalog No. 50-004-08042 Supplier Medchemexpress LLC Supplier No. HYB0782R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetazolamide (Standard) is an analytical standard of Acetazolamide, designed for research and analytical applications. It functions as a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX and possesses diuretic, antihypertensive, and anti-gonococcal properties. This product is specifically intended for laboratory use and is not sold to patients.

  • Analytical standard for assays
  • Intended for qualitative, quantitative, and methodological research experiments
  • Commonly used in chromatography (HPLC, GC, MS)
  • High purity of 99.69%
  • Appears as a white to off-white solid
  • Molecular weight: 222.25
  • Chemical formula: C₄H₆N₄O₃S₂
  • Not for human use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.