Learn More
Medchemexpress LLC Acetazolamide (Standard) | 59-66-5 | 99.9% | 5 G
SDP
Catalog No. 5000407734
Encompass_Preferred
Acetazolamide is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It also exhibits diuretic, antihypertensive, and anti-gonococcal activities, and inhibits hCA II with an IC50 of 130 nM. This small heteroaromatic sulfonamide binds to various carbonic anhydrases with high affinity.
  • Carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX
  • Diuretic, antihypertensive, and anti-gonococcal activities
  • Inhibits hCA II with an IC50 of 130 nM
  • Small heteroaromatic sulfonamide
  • Binds to various carbonic anhydrases with high affinity
Catalog No. 50-004-07734 Supplier Medchemexpress LLC Supplier No. HYB07825G
Acetazolamide is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It also exhibits diuretic, antihypertensive, and anti-gonococcal activities, and inhibits hCA II with an IC50 of 130 nM. This small heteroaromatic sulfonamide binds to various carbonic anhydrases with high affinity.
  • Carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX
  • Diuretic, antihypertensive, and anti-gonococcal activities
  • Inhibits hCA II with an IC50 of 130 nM
  • Small heteroaromatic sulfonamide
  • Binds to various carbonic anhydrases with high affinity