Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetazolamide, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18600201 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-201 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-600-201 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A12665KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A1266, 59-66-5, C4H6N4O3S2

Spectrum™ Chemical Acetazolamide, USP is in the diuretic and carbonic anhydrase inhibitor families of medications. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 59-66-5
CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Pail
Molecular Formula C4H6N4O3S2
MDL Number MFCD00003105
Quantity 5 kg
InChI Key BZKPWHYZMXOIDC-UHFFFAOYSA-N
SMILES CC(=O)NC1=NN=C(S1)S(N)(=O)=O
IUPAC Name N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
Molecular Weight (g/mol) 222.24
Percent Purity 98 to 102%
Grade USP
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.