Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetc acd glacal ACS re500ML
Catalog No. 502903587
Encompass
Acetic acid is an organic carboxylic acid. It is a pungent smelling hygroscopic liquid. Glacial acetic acid is the anhydrous acetic acid. It forms dimers linked by hydrogen bonds in SOLUTIONon and gaseous commonly referred as phases. It is weakly acidic in nature.
Catalog No. 50-290-3587 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6950926X500MLGL
Acetic acid is an organic carboxylic acid. It is a pungent smelling hygroscopic liquid. Glacial acetic acid is the anhydrous acetic acid. It forms dimers linked by hydrogen bonds in SOLUTIONon and gaseous commonly referred as phases. It is weakly acidic in nature.