Learn More
Electron Microscopy Sciences Acetic Acid 0.2% Aqueous 500 ML
SDP
Catalog No. 5030056
Encompass_Preferred
Used in Price's Giemsa Stain, for hematologic elements. See references AFIP Manual of Histological Staining Methods, 3rd ed., L. Luna: New York: McGraw Hill Publications, c. 1968, p. 127 and Price, D.L., Mil. Med., 133: 363-367, 1968.
Catalog No. 50-300-56 Supplier Electron Microscopy Sciences Supplier No. 2615603
Used in Price's Giemsa Stain, for hematologic elements. See references AFIP Manual of Histological Staining Methods, 3rd ed., L. Luna: New York: McGraw Hill Publications, c. 1968, p. 127 and Price, D.L., Mil. Med., 133: 363-367, 1968.