Learn More
Electron Microscopy Sciences Acetic Acid 0.2% Aqueous 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2615503
Encompass_Preferred
Used in Thomas Method for Malarial Parasites, Thomas (1953). References: Thomas, J.T. Stain Techn., 28:311-312, 1953 and Luna, L.G. (ed.) Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, New York, P.129, c.1968
Catalog No. 50-317-06