Learn More
Electron Microscopy Sciences Acetic Acid 1% 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2661304
Encompass_Preferred
Used in Prussian Blue Method for Hemosiderin for the staining of deposits of hemosiderin. For references see Perls, M., Virchow's Arch. F. Path. Anat., 39:42-48, 1867, Highman, B., Arch. Path., 33:937-938, 1942, Lillie, R.D. Histopathologic Technic and Practical Histo-chemistry, 3rd Ed. McGraw-Hill, N.Y., 1965 or Clark, G., (ed), Staining Procedures 3rd Ed., Williams & Wilkins, Baltimore, p. 157, c. 1973.
Catalog No. 50-318-68