Learn More
Electron Microscopy Sciences Acetic Acid 1% Aqueous 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2636707
Encompass_Preferred
Used in Masson's Trichrome for Connective Tissue for stain nuclei, agentaffen granules, collagen, cytoplasm, keratin, etc. References: Sheehan and Hrapchak: Theory and Practice of Histotechnology, St. Louis. The Moxby Company 1980, p. 190
Catalog No. 50-317-78