Learn More
Research Products International Corp Acetic Acid, Glacial, 1 Liter, Research or Further Manufacturing Use Only, Not for Food or Drug Use
SDP
Catalog No. 501977981
Encompass_Preferred
Acetic acid (CH₃COOH) is a colorless, corrosive liquid with a strong odor, fully miscible in water. With a molecular weight of 60.05 g/mol, it is used in life science labs for buffer preparation, protein precipitation, histological staining, pH regulation, and as a solvent in various biochemical and molecular procedures.
Catalog No. 50-197-7981 Supplier Research Products International Corp Supplier No. A128001000.0
Acetic acid (CH₃COOH) is a colorless, corrosive liquid with a strong odor, fully miscible in water. With a molecular weight of 60.05 g/mol, it is used in life science labs for buffer preparation, protein precipitation, histological staining, pH regulation, and as a solvent in various biochemical and molecular procedures.