Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid 100%, For LC-MS LiChropur™, MilliporeSigma™

Catalog No. 01100263 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-100-263 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-100-263 Supplier MilliporeSigma Supplier No. 5.33001.0050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Ethanoic acid
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5 (50 g/L water at 20°C)
Melting Point 16.64°C
Boiling Point 116°C to 118°C at 1013 hPa
Density 1.04 g/cm3 at 25 °C
Quantity 50 mL
Explosion Limit 4 to 19.9% (V)
Flash Point 40 °C
Packaging Glass bottle
Vapor Pressure 20.79 hPa at 25 °C
Solubility Information 602.9 g/L
Recommended Storage Store at 15 to 25 °C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.