Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid (100%, Glacial), Anhydrous, EMSURE™ MilliporeSigma™

Catalog No. M1000639026 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Glass bottle
Plastic bottle
Safebreak bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1000639026 25 L Plastic bottle
M1000632500 2.5 L Glass bottle
M1000632511 2.5 L Plastic bottle
M1000632510 2.5 L Safebreak bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1000639026 Supplier MilliporeSigma Supplier No. 1.00063.9026
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • 100% anhydrous for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5 (50g/L, H2O, 20°C)
Melting Point 17°C
Boiling Point 16°C to 118°C (1013hPa)
Color Colorless
Density 1.05g/cm3
Quantity 25 L
Explosion Limit 4 to 19.9%
Flash Point 39°C
Ignition Point 485°C
Odor Pungent/Vinegar-like
Packaging Plastic bottle
Physical Form Liquid
Residue after Evaporation <5ppm
Vapor Pressure 15.4hPa(20°C)
UN Number UN 2789
Solubility Information 602.9g/L
Formula Weight 60.05g/mol
Grade ACS/ISO/Reagent Ph. Eur.
Chemical Name or Material Acetic Acid
Show More Show Less
Hazard Category Corrosive
Hazard Statement H226: Flammable liquid and vapor, H290: May be corrosive to metals, H314: Causes severe skin burns and eye damage,
Precautionary Statement P210: Keep away from heat, P280: Wear protective gloves/ protective clothing/ eye protection/ face protection, P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting
EINECSNumber 200-580-7
RTECSNumber AF1225000
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.