Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid 100%, MilliporeSigma

Catalog No. 01362683 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-362-683 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-362-683 Supplier MilliporeSigma Supelco Supplier No. 9683650ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tested for PFAS methods, LiChropur.

  • Suitable for EPA 1633, EPA 533, EPA 537.1
  • HPLC, LC/MS, ion chromatography suitable
TRUSTED_SUSTAINABILITY

Specifications

CAS 64-19-7
Quantity 50 mL
Molecular Weight (g/mol) 60.05
Percent Purity ≥99.8% (Acidimetric)
Chemical Name or Material Acetic Acid 100%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.