Learn More
Medchemexpress LLC Acetic acid, 2-[(18-chloro-3,6,9,12-tetraoxaoctadec-1-yl)oxy]- | 1799506-30-1 | 98.0% | 370.87 | 250 MG
SDP
Catalog No. 5000439158
Encompass_Preferred
Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is a PEG-based PROTAC linker designed for the synthesis of chloroalkane-containing PROTACs, also known as HaloPROTACs. PROTACs function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs).
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system.
Catalog No. 50-004-39158 Supplier Medchemexpress LLC Supplier No. 250MGHY112496
Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is a PEG-based PROTAC linker designed for the synthesis of chloroalkane-containing PROTACs, also known as HaloPROTACs. PROTACs function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs).
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system.