Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetic acid, 2-[2-[2-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino] | 2097938-44-6 | 99.2% | 507.49 | 25 MG
SDP

Catalog No. 5000402193
Encompass_Preferred

Pomalidomide-PEG4-C-COOH (Cereblon Ligand -Linker Conjugate 1) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker used in PROTAC technology. It is for research use only.

  • Purity of 99.19%
  • Molecular weight of 507.49
  • Chemical formula: C23H29N3O10
  • Appearance as a light yellow to yellow oil
  • Soluble in DMSO and ethanol
  • Store at 4°C under nitrogen
  • Target: Cereblon
  • Utilized in PROTAC technology to selectively degrade target proteins

Catalog No. 50-004-02193 Supplier Medchemexpress LLC Supplier No. 25MGHY21930
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-PEG4-C-COOH (Cereblon Ligand -Linker Conjugate 1) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker used in PROTAC technology. It is for research use only.

  • Purity of 99.19%
  • Molecular weight of 507.49
  • Chemical formula: C23H29N3O10
  • Appearance as a light yellow to yellow oil
  • Soluble in DMSO and ethanol
  • Store at 4°C under nitrogen
  • Target: Cereblon
  • Utilized in PROTAC technology to selectively degrade target proteins

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.