Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetic acid, 2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]-, 1,1-dimethylethyl ester | 189808-70-6 | ≥98.0% | 250 MG
SDP

Catalog No. 5000341480
Encompass_Preferred

NH2-PEG3-C1-Boc, also known as PROTAC Linker 5, is a PEG-based PROTAC linker. It is primarily used in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Purity of at least 98.0%
  • Appearance is a colorless to light yellow liquid
  • Molecular formula is C12H25NO5
  • Molecular weight is 263.33
  • CAS number is 189808-70-6
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-41480 Supplier Medchemexpress LLC Supplier No. 250MGHY128801
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-PEG3-C1-Boc, also known as PROTAC Linker 5, is a PEG-based PROTAC linker. It is primarily used in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Purity of at least 98.0%
  • Appearance is a colorless to light yellow liquid
  • Molecular formula is C12H25NO5
  • Molecular weight is 263.33
  • CAS number is 189808-70-6
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.