Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetic acid, 2-[2-[2-[[(4-methylphenyl)sulfonyl)oxy]ethoxy]ethoxy]-, 1,1-dimethylethyl ester | 882518-89-0 | 97.0% | 25 MG
SDP

Catalog No. 5000341745
Encompass_Preferred

This product is Acetic acid, 2-[2-[2-[[(4-methylphenyl)sulfonyl)oxy]ethoxy]ethoxy]-, 1,1-dimethylethyl ester, with a CAS number of 882518-89-0. It has a molecular formula of C17H26O7S and a molecular weight of 374.45. This colorless to light yellow liquid has a purity of 97.0%.

  • Colorless to light yellow liquid
  • Purity of 97.0%
  • Molecular formula: C17H26O7S
  • Molecular weight: 374.45
  • Store at -20°C, protected from light

Catalog No. 50-003-41745 Supplier Medchemexpress LLC Supplier No. 25MGHY130532
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is Acetic acid, 2-[2-[2-[[(4-methylphenyl)sulfonyl)oxy]ethoxy]ethoxy]-, 1,1-dimethylethyl ester, with a CAS number of 882518-89-0. It has a molecular formula of C17H26O7S and a molecular weight of 374.45. This colorless to light yellow liquid has a purity of 97.0%.

  • Colorless to light yellow liquid
  • Purity of 97.0%
  • Molecular formula: C17H26O7S
  • Molecular weight: 374.45
  • Store at -20°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.