Learn More
Medchemexpress LLC Acetic acid, 2-[2-(2-aminoethoxy)ethoxy]- | 134978-97-5 | 163.17 | 5 G
SDP
Catalog No. 5000446207
Encompass_Preferred
H2N-PEG2-CH2COOH is a PEG-based PROTAC linker, appearing as a white to yellow solid. This compound can be utilized in the synthesis of PROTACs, which are molecular tools composed of two distinct ligands joined by a linker. One ligand binds to an E3 ubiquitin ligase, and the other targets a protein of interest, allowing PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • White to yellow solid appearance
  • High purity (99.87%)
Catalog No. 50-004-46207 Supplier Medchemexpress LLC Supplier No. HYW0065245G
H2N-PEG2-CH2COOH is a PEG-based PROTAC linker, appearing as a white to yellow solid. This compound can be utilized in the synthesis of PROTACs, which are molecular tools composed of two distinct ligands joined by a linker. One ligand binds to an E3 ubiquitin ligase, and the other targets a protein of interest, allowing PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • White to yellow solid appearance
  • High purity (99.87%)