Learn More
Medchemexpress LLC Acetic acid, 2-[2-(2-azidoethoxy)ethoxy]- | 882518-90-3 | 99.0% | 189.17 | 1 MG
SDP
Catalog No. 5000346871
Encompass_Preferred
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Can be used in the synthesis of PROTACs
  • PROTACs utilize the intracellular ubiquitin-proteasome system to degrade target proteins
Catalog No. 50-003-46871 Supplier Medchemexpress LLC Supplier No. HY10836810G
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Can be used in the synthesis of PROTACs
  • PROTACs utilize the intracellular ubiquitin-proteasome system to degrade target proteins