Learn More
Medchemexpress LLC Acetic acid, 2-[2-(2-azidoethoxy)ethoxy]- | 882518-90-3 | 99.03% | 189.17 | 10 MG
SDP
Catalog No. 5000347122
Encompass_Preferred
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Can be used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Azide group enables copper-catalyzed azide-alkyne cycloaddition.
  • Can undergo strain-promoted alkyne-azide cycloaddition with DBCO or BCN groups.
Catalog No. 50-003-47122 Supplier Medchemexpress LLC Supplier No. HY108368A100MG
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Can be used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Azide group enables copper-catalyzed azide-alkyne cycloaddition.
  • Can undergo strain-promoted alkyne-azide cycloaddition with DBCO or BCN groups.