Learn More
Medchemexpress LLC Acetic acid, 2-[2-(2-azidoethoxy)ethoxy]- | 882518-90-3 | 99.03% | 189.17 | 50 MG
SDP
Catalog No. 5000347139
Encompass_Preferred
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This product is for research use only.
  • Peg-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group for CuAAc and SPAAC reactions
Catalog No. 50-003-47139 Supplier Medchemexpress LLC Supplier No. HY1083681G
Add to Cart
Add to Cart
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This product is for research use only.
  • Peg-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group for CuAAc and SPAAC reactions