Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetic acid, 2-(2,4-dichlorophenoxy)-, butyl ester | 94-80-4 | 99.9% | 1 G
SDP

Catalog No. 5000407657
Encompass_Preferred

2,4-D Butyl ester is an organic phenoxy herbicide used to control woody broad-leaf weeds. It functions by mimicking natural plant growth hormones, causing plants to grow too rapidly for survival.

  • Acts as a herbicide
  • Targets metabolic enzyme/protease pathway
  • Appears as a colorless to light yellow liquid
  • Soluble in DMSO at 100 mg/mL
  • Mimics natural plant growth hormones
  • Used to control woody broad-leaf weeds
  • Molecular formula: C12H14Cl2O3
  • Molecular weight: 277.14
  • Purity: 99.9%

Catalog No. 50-004-07657 Supplier Medchemexpress LLC Supplier No. HYB08671G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4-D Butyl ester is an organic phenoxy herbicide used to control woody broad-leaf weeds. It functions by mimicking natural plant growth hormones, causing plants to grow too rapidly for survival.

  • Acts as a herbicide
  • Targets metabolic enzyme/protease pathway
  • Appears as a colorless to light yellow liquid
  • Soluble in DMSO at 100 mg/mL
  • Mimics natural plant growth hormones
  • Used to control woody broad-leaf weeds
  • Molecular formula: C12H14Cl2O3
  • Molecular weight: 277.14
  • Purity: 99.9%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.