Learn More
Medchemexpress LLC Acetic acid, 2-(2,4-dichlorophenoxy)-, sodium salt (1:1) | 2702-72-9 | ≥98.0% | 243.02 | 10 G
SDP
Catalog No. 5000387856
Encompass_Preferred
2,4-D sodium is a selective herbicide primarily used for the control of broad-leaved weeds. This compound induces apoptosis and inhibits DNA and protein synthesis, which prevents normal plant growth and development. Research indicates it may also have toxic effects on the reproductive system in male rats at specific dosages.
  • Selective herbicide for broad-leaved weeds.
  • Induces apoptosis.
  • Inhibits DNA and protein synthesis.
  • Prevents normal plant growth and development.
Catalog No. 50-003-87856 Supplier Medchemexpress LLC Supplier No. HY18572A10G
Add to Cart
Add to Cart
2,4-D sodium is a selective herbicide primarily used for the control of broad-leaved weeds. This compound induces apoptosis and inhibits DNA and protein synthesis, which prevents normal plant growth and development. Research indicates it may also have toxic effects on the reproductive system in male rats at specific dosages.
  • Selective herbicide for broad-leaved weeds.
  • Induces apoptosis.
  • Inhibits DNA and protein synthesis.
  • Prevents normal plant growth and development.