Learn More
Medchemexpress LLC Acetic acid, 2-(2-azidoethoxy)- | 79598-48-4 | 99.6% | 145.12 | 250 MG
SDP
Catalog No. 5000347756
Encompass_Preferred
Azido-PEG1-CH2CO2H is a PROTAC linker, characterized by its alkyl/ether composition. It is utilized in the synthesis of PROTAC BRD4 Degrader-1. This compound also functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PROTAC linker with alkyl/ether composition
  • Used in the synthesis of PROTAC BRD4 Degrader-1
  • Functions as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • For research use only
Catalog No. 50-003-47756 Supplier Medchemexpress LLC Supplier No. 250MGHY108369
Azido-PEG1-CH2CO2H is a PROTAC linker, characterized by its alkyl/ether composition. It is utilized in the synthesis of PROTAC BRD4 Degrader-1. This compound also functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PROTAC linker with alkyl/ether composition
  • Used in the synthesis of PROTAC BRD4 Degrader-1
  • Functions as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • For research use only