Learn More
Medchemexpress LLC Acetic acid, 2-(2-cyclooctyn-1-yloxy)-, 2,5-dioxo-1-pyrrolidinyl ester | 1425803-45-7 | MFCD28898688 | >95.0% | 279.29 | C14H17NO5 | 5 MG
SDP
Catalog No. 5000201879
Encompass_Preferred
Cyclooctyne-O-NHS ester is an NHS-activated cyclooctyne reagent used as a cleavable linker and click-chemistry reagent for in vitro bioconjugation and antibody-drug conjugate synthesis. The molecule combines a strained alkyne for azide-alkyne cycloaddition with an activated ester to enable efficient amine coupling.
  • Enables azide-alkyne cycloaddition reactions for bioorthogonal conjugation.
  • Activated NHS ester facilitates rapid coupling to primary amines.
  • Designed for use as a cleavable linker in ADC synthesis and related conjugation workflows.
  • Provided in small, laboratory-scale quantities suitable for research use.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for traceability and safety.
Catalog No. 50-002-01879 Supplier Medchemexpress LLC Supplier No. HY1265175MG
Cyclooctyne-O-NHS ester is an NHS-activated cyclooctyne reagent used as a cleavable linker and click-chemistry reagent for in vitro bioconjugation and antibody-drug conjugate synthesis. The molecule combines a strained alkyne for azide-alkyne cycloaddition with an activated ester to enable efficient amine coupling.
  • Enables azide-alkyne cycloaddition reactions for bioorthogonal conjugation.
  • Activated NHS ester facilitates rapid coupling to primary amines.
  • Designed for use as a cleavable linker in ADC synthesis and related conjugation workflows.
  • Provided in small, laboratory-scale quantities suitable for research use.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for traceability and safety.