Learn More
Medchemexpress LLC Acetic acid, 2-(2-cyclooctyn-1-yloxy)-, 2,5-dioxo-1-pyrrolidinyl ester | 1425803-45-7 | MFCD28898688 | 95.0% | 279.29 g/mol | C14H17NO5 | 50 MG
SDP
Catalog No. 5000201887
Encompass_Preferred
Cyclooctyne-O-NHS ester is a cyclooctyne-containing N-hydroxysuccinimide (NHS) ester used as a cleavable ADC linker and click-chemistry reagent for bioconjugation. It undergoes azide-alkyne cycloaddition to enable antibody-drug conjugate construction, labeling, and related conjugation workflows.
  • Cleavable ADC linker for antibody-drug conjugate synthesis.
  • Reacts with azide groups via strain-promoted and copper-catalyzed azide-alkyne cycloaddition.
  • NHS ester chemistry enables efficient conjugation to primary amines.
  • Supplied as a solid in multiple pack sizes suitable for small-scale synthesis.
  • Datasheet and safety data sheet available for handling and storage guidance.
Catalog No. 50-002-01887 Supplier Medchemexpress LLC Supplier No. HY12651750MG
Cyclooctyne-O-NHS ester is a cyclooctyne-containing N-hydroxysuccinimide (NHS) ester used as a cleavable ADC linker and click-chemistry reagent for bioconjugation. It undergoes azide-alkyne cycloaddition to enable antibody-drug conjugate construction, labeling, and related conjugation workflows.
  • Cleavable ADC linker for antibody-drug conjugate synthesis.
  • Reacts with azide groups via strain-promoted and copper-catalyzed azide-alkyne cycloaddition.
  • NHS ester chemistry enables efficient conjugation to primary amines.
  • Supplied as a solid in multiple pack sizes suitable for small-scale synthesis.
  • Datasheet and safety data sheet available for handling and storage guidance.