Learn More
Medchemexpress LLC Acetic acid, 2,2-dihydroxy- | 563-96-2 | MFCD00127974 | 98.0% | 92.05 | C2H4O4 | 10 G
SDP
Catalog No. 5000301489
Encompass_Preferred
2,2-Dihydroxyacetic acid is a small-molecule chemical intermediate and reagent used in organic synthesis and analytical assays. It serves as a building block for the preparation of 3-aminocoumarone derivatives and is applied in chemiluminescence-based detection of hydrogen peroxide and in enzymatic activity evaluations.
  • High purity (98.0%).
  • Solid, white to light yellow appearance for easy handling.
  • Molecular formula C2H4O4 and molecular weight 92.05 g·mol⁻¹.
  • Available in multiple pack sizes, including 10 g, for flexible use.
  • Suitable as a synthesis intermediate and analytical reagent.
Catalog No. 50-003-01489 Supplier Medchemexpress LLC Supplier No. HYW01972410G
Add to Cart
Add to Cart
2,2-Dihydroxyacetic acid is a small-molecule chemical intermediate and reagent used in organic synthesis and analytical assays. It serves as a building block for the preparation of 3-aminocoumarone derivatives and is applied in chemiluminescence-based detection of hydrogen peroxide and in enzymatic activity evaluations.
  • High purity (98.0%).
  • Solid, white to light yellow appearance for easy handling.
  • Molecular formula C2H4O4 and molecular weight 92.05 g·mol⁻¹.
  • Available in multiple pack sizes, including 10 g, for flexible use.
  • Suitable as a synthesis intermediate and analytical reagent.