Learn More
Medchemexpress LLC Acetic acid, 2-[[4-(3-methoxyphenyl)-5-(2-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]-, sodium salt | 1459687-96-7 | 99.5% | 364.35 g/mol | C16H13N4NaO3S | 1 ML
SDP
Catalog No. 5000212884
Encompass_Preferred
GJ103 sodium is the sodium salt form of a small-molecule read-through compound that promotes translational read-through of premature stop codons. It is provided for research use in studies of genetic disorders caused by nonsense mutations and is available as a high-purity solid or as a concentrated solution in DMSO.
  • Induces read-through of premature stop codons for genetic research.
  • Water-soluble sodium salt form enables easier handling and formulation.
  • High purity (reported ~99.5%) suitable for research applications.
  • Offered as a 10 mM solution in DMSO and in multiple solid mass sizes.
  • Molecular weight approximately 364.35 g/mol.
Catalog No. 50-002-12884 Supplier Medchemexpress LLC Supplier No. HY101203A10MM 1ML
GJ103 sodium is the sodium salt form of a small-molecule read-through compound that promotes translational read-through of premature stop codons. It is provided for research use in studies of genetic disorders caused by nonsense mutations and is available as a high-purity solid or as a concentrated solution in DMSO.
  • Induces read-through of premature stop codons for genetic research.
  • Water-soluble sodium salt form enables easier handling and formulation.
  • High purity (reported ~99.5%) suitable for research applications.
  • Offered as a 10 mM solution in DMSO and in multiple solid mass sizes.
  • Molecular weight approximately 364.35 g/mol.