Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetic acid, 2-[4-[[4-(2-chlorophenyl)-2-thiazolyl]methoxy]-2-methylphenoxy]- | 1050506-75-6 | 98.1% | C19H16ClNO4S | 1 ML
SDP

Catalog No. 5000359210
Encompass_Preferred

GPR120 modulator 1 is a G protein coupled receptor 120 (GPR120) modulator. This compound, extracted from patent US8394841B2 (compound example F1), is suitable for research into diseases linked to abnormal or deregulated GPR120, including diabetes.

  • Modulates G protein coupled receptor 120 (GPR120)
  • Supports research on diseases linked to GPR120 deregulation, such as diabetes
  • Purity of 98.1% (HPLC)
  • White to off-white solid appearance
  • Chemical formula C19H16ClNO4S

Catalog No. 50-003-59210 Supplier Medchemexpress LLC Supplier No. HY5016210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GPR120 modulator 1 is a G protein coupled receptor 120 (GPR120) modulator. This compound, extracted from patent US8394841B2 (compound example F1), is suitable for research into diseases linked to abnormal or deregulated GPR120, including diabetes.

  • Modulates G protein coupled receptor 120 (GPR120)
  • Supports research on diseases linked to GPR120 deregulation, such as diabetes
  • Purity of 98.1% (HPLC)
  • White to off-white solid appearance
  • Chemical formula C19H16ClNO4S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.