Learn More
Medchemexpress LLC Acetic acid, [2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl-2,2,3,3,5,5,6,6-d8]ethoxy]- | 774596-22-4 | 96.5% | 396.94 | 1 MG
SDP
Catalog No. 5000384304
Encompass_Preferred
Cetirizine-d8 is a deuterium labeled Cetirizine. It is a specific, orally active, and long-acting histamine H1-receptor antagonist with antiallergic properties, inhibiting eosinophil chemotaxis during the allergic response.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-84304 Supplier Medchemexpress LLC Supplier No. HY17042S11MG
Cetirizine-d8 is a deuterium labeled Cetirizine. It is a specific, orally active, and long-acting histamine H1-receptor antagonist with antiallergic properties, inhibiting eosinophil chemotaxis during the allergic response.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS