Learn More
Medchemexpress LLC Acetic acid, 2-[[8-chloro-3-[(4-chlorophenyl)methyl]-4-(difluoromethoxy)-2-ethyl-5-quinolinyl... | 932708-14-0 | 99.4% | 456.27 | 50 MG
SDP
Catalog No. 5000345178
Encompass_Preferred
Pexopiprant is an oral antagonist of the prostaglandin D2 receptor 2 (DP2) with a Ki of less than 100 nM. It can be used in studies of asthma.
  • Oral antagonist of DP2
  • Ki < 100 nM
  • Potential use in asthma studies
  • For research use only
  • Available in solid and solution forms
Catalog No. 50-003-45178 Supplier Medchemexpress LLC Supplier No. HY10918650MG
Pexopiprant is an oral antagonist of the prostaglandin D2 receptor 2 (DP2) with a Ki of less than 100 nM. It can be used in studies of asthma.
  • Oral antagonist of DP2
  • Ki < 100 nM
  • Potential use in asthma studies
  • For research use only
  • Available in solid and solution forms