Learn More
Medchemexpress LLC Acetic acid, 2-azido- | 18523-48-3 | C2H3N3O2 | 250 MG
SDP
Catalog No. 5000374477
Encompass_Preferred
Azidoacetic acid is a click chemistry reagent containing an azide group. It functions as a small molecule tool for the synthesis of PROTACs. The azide group enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Contains an azide group
  • Used as a small molecule tool for PROTAC synthesis
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Click chemistry reagent for selective, modular, high-yield reactions
  • Supports bioorthogonal chemistry for reactions that do not interfere with biological processes
Catalog No. 50-003-74477 Supplier Medchemexpress LLC Supplier No. 250MGHY151719
Azidoacetic acid is a click chemistry reagent containing an azide group. It functions as a small molecule tool for the synthesis of PROTACs. The azide group enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Contains an azide group
  • Used as a small molecule tool for PROTAC synthesis
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Click chemistry reagent for selective, modular, high-yield reactions
  • Supports bioorthogonal chemistry for reactions that do not interfere with biological processes