Learn More
Medchemexpress LLC Acetic acid, 2-bromo-, 2,5-dioxo-1-pyrrolidinyl ester | 42014-51-7 | 99.2% | 236.02 | 1 G
SDP
Catalog No. 5000452990
Encompass_Preferred
N-Succinimidyl bromoacetate is a heterobifunctional cross-linking reagent that can be used to prepare peptide-protein immunogens.
  • Acts as a heterobifunctional cross-linking reagent.
  • Used to prepare peptide-protein immunogens.
Catalog No. 50-004-52990 Supplier Medchemexpress LLC Supplier No. HYW0121661G
N-Succinimidyl bromoacetate is a heterobifunctional cross-linking reagent that can be used to prepare peptide-protein immunogens.
  • Acts as a heterobifunctional cross-linking reagent.
  • Used to prepare peptide-protein immunogens.