Learn More
Medchemexpress LLC Acetic acid, 2-cyano-, 2-[[(3,4-dichlorophenyl)amino]thioxomethyl] hydrazide | 656222-54-7 | 99.8% | 303.17 g/mol | C10H8Cl2N4OS | 10MG
SDP
Catalog No. 5000239299
Encompass_Preferred
iKIX1 is a small-molecule antifungal research compound (CAS 656222-54-7) that inhibits the interaction between the KIX domain of the mediator subunit CgGal11A and the activation domain of CgPdr1, resensitizing drug-resistant Candida glabrata to azole antifungals in vitro. It is supplied for research use and is not for human or clinical use.
  • Resensitizes azole-resistant Candida glabrata in vitro.
  • Inhibits CgGal11A-CgPdr1 interaction to reduce multidrug resistance.
  • High purity suitable for research (HPLC purity 99.76%).
  • Molecular formula C10H8Cl2N4OS; molecular weight 303.17 g/mol.
  • Includes certificate of analysis and safety data sheet for quality and handling information.
Catalog No. 50-002-39299 Supplier Medchemexpress LLC Supplier No. HY12495210MG
iKIX1 is a small-molecule antifungal research compound (CAS 656222-54-7) that inhibits the interaction between the KIX domain of the mediator subunit CgGal11A and the activation domain of CgPdr1, resensitizing drug-resistant Candida glabrata to azole antifungals in vitro. It is supplied for research use and is not for human or clinical use.
  • Resensitizes azole-resistant Candida glabrata in vitro.
  • Inhibits CgGal11A-CgPdr1 interaction to reduce multidrug resistance.
  • High purity suitable for research (HPLC purity 99.76%).
  • Molecular formula C10H8Cl2N4OS; molecular weight 303.17 g/mol.
  • Includes certificate of analysis and safety data sheet for quality and handling information.