Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetic acid, 2-iodo-, 1,1'-anhydride | 54907-61-8 | MFCD00001080 | 70.0% | 353.88 | C4H4I2O3 | 50 MG
SDP

Catalog No. 5000284165
Encompass_Preferred

Iodoacetic anhydride is an acylating reagent used in organic synthesis and biochemical research for selective modification of thiol groups and protein labelling. It is supplied as a brown to dark brown solid and should be handled under anhydrous conditions with appropriate cold storage to preserve stability.

  • Acylating reagent for protein modification and organic synthesis.
  • Contains 2-iodoacetyl functionality for selective alkylation of thiols.
  • Provided as a brown to dark brown solid suitable for analytical use.
  • Reported purity: 70.0%.
  • Recommended storage: cold, anhydrous conditions to maintain stability.
  • Handle in a fume hood with appropriate personal protective equipment.

Catalog No. 50-002-84165 Supplier Medchemexpress LLC Supplier No. HYW68660550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iodoacetic anhydride is an acylating reagent used in organic synthesis and biochemical research for selective modification of thiol groups and protein labelling. It is supplied as a brown to dark brown solid and should be handled under anhydrous conditions with appropriate cold storage to preserve stability.

  • Acylating reagent for protein modification and organic synthesis.
  • Contains 2-iodoacetyl functionality for selective alkylation of thiols.
  • Provided as a brown to dark brown solid suitable for analytical use.
  • Reported purity: 70.0%.
  • Recommended storage: cold, anhydrous conditions to maintain stability.
  • Handle in a fume hood with appropriate personal protective equipment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.