Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetic acid, 2-iodo-, 1,1'-anhydride | 54907-61-8 | MFCD00001080 | 70.0% | 353.88 g·mol-1 | C4H4I2O3 | 100 MG
SDP

Catalog No. 5000284517
Encompass_Preferred

Iodoacetic anhydride is the 2-iodo derivative of acetic anhydride used as a reagent in chemical synthesis and biochemical research. It is supplied as a brown to dark brown solid with molecular formula C4H4I2O3 and molecular weight 353.88 g·mol-1.

  • Molecular formula: C4H4I2O3
  • Molecular weight: 353.88 g·mol-1
  • Appearance: Brown to dark brown solid
  • Cas number: 54907-61-8
  • Typical pack size: 100 MG
  • Purity (batch example): 70.0% (NMR)

Catalog No. 50-002-84517 Supplier Medchemexpress LLC Supplier No. HYW686605100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iodoacetic anhydride is the 2-iodo derivative of acetic anhydride used as a reagent in chemical synthesis and biochemical research. It is supplied as a brown to dark brown solid with molecular formula C4H4I2O3 and molecular weight 353.88 g·mol-1.

  • Molecular formula: C4H4I2O3
  • Molecular weight: 353.88 g·mol-1
  • Appearance: Brown to dark brown solid
  • Cas number: 54907-61-8
  • Typical pack size: 100 MG
  • Purity (batch example): 70.0% (NMR)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.