Learn More
Medchemexpress LLC Acetic acid, 2-iodo-, sodium salt | 305-53-3 | 99.9% | 10 G
SDP
Catalog No. 5000410436
Encompass_Preferred
Sodium iodoacetate is a specific inhibitor of GAPDH and exhibits glycolysis inhibitory activity. It can also induce osteoarthritis and related pain models in experimental animals.
  • Specific inhibitor of GAPDH.
  • Has glycolysis inhibitory activity.
  • Can induce osteoarthritis and related pain models in experimental animals.
  • Useful for research involving GAPDH inhibition.
  • Applicable in studies related to glycolysis inhibition.
  • Can be used to establish osteoarthritis and pain models in experimental animals.
Catalog No. 50-004-10436 Supplier Medchemexpress LLC Supplier No. HYD084910G
Sodium iodoacetate is a specific inhibitor of GAPDH and exhibits glycolysis inhibitory activity. It can also induce osteoarthritis and related pain models in experimental animals.
  • Specific inhibitor of GAPDH.
  • Has glycolysis inhibitory activity.
  • Can induce osteoarthritis and related pain models in experimental animals.
  • Useful for research involving GAPDH inhibition.
  • Applicable in studies related to glycolysis inhibition.
  • Can be used to establish osteoarthritis and pain models in experimental animals.