Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, 5% (v/v), Ricca Chemical
BIOPHARMASDP

Catalog No. 13016
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5, 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 94.0, 4.0
Color Colorless
CAS Max % 96.0, 6.0
Appearance Colorless Liquid
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
Concentration 5% (v/v)
Percent Purity 4.94 to 5.06%
Grade Laboratory
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Acetic Acid, 5% (v/v)
Show More Show Less
ShelfLife 36 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.