Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMarKa Chemical Company Acetic Acid, 50% V/V, 4l

Catalog No. 504400002
Encompass_Preferred

Acetic Acid, 64-49-7, WT%: 50%, OSHA/PEL: 10 PPM, ACGIH/TLV: 10PPM, 4l

Catalog No. 50-440-0002 Supplier LaMarKa Chemical Company Supplier No. 00000012167532436
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetic Acid, 64-49-7, WT%: 50%, OSHA/PEL: 10 PPM, ACGIH/TLV: 10PPM, 4l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.