Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid Standard, 500 ppm CH3COOH in 10% (v/v) Ethanol, Ricca Chemical
SDP

Catalog No. r0091500100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0091500100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R0091500100 Supplier Ricca Chemical Company Supplier No. R0091500100A
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-19-7 , 7732-18-5 , 64-17-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 0.03, 89.0, 9.0
CAS Max % 0.03, 91.0, 11.0
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Acetic Acid Standard, 500ppm in 10% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.