Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid 96%, Supelco

Catalog No. 11100259 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-259 2.5 L
11-100-258 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-259 Supplier MilliporeSigma Supelco Supplier No. 1000622511

missing translation for 'validMainframeMsg'

For analysis EMSURE™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Weight (g/mol) 60.05

Specifications

Percent Purity Unknown
Grade Ungraded
Quantity 2.5 L
Packaging HDPE Bottle
Chemical Name or Material Acetic Acid 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.