Learn More
Sigma Aldrich Fine Chemicals Biosciences Glacial Acetic acid, 64-19-7, MFCD00036152, 500mL
Catalog No. 501657098
Encompass
Linear Formula CH3CO2H, Purity ≥99.7%, Molecular Weight 60.05, ACS reagent, Acetic acid is an organic carboxylic acid. It is a pungent smelling hygroscopic liquid. Glacial acetic acid is the anhydrous acetic acid. It forms dimers linked by hydrogen bonds in solution and gaseous commonly referred as phases. It is weakly acidic in nature.
Catalog No. 50-165-7098 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 695092500MLGL
Linear Formula CH3CO2H, Purity ≥99.7%, Molecular Weight 60.05, ACS reagent, Acetic acid is an organic carboxylic acid. It is a pungent smelling hygroscopic liquid. Glacial acetic acid is the anhydrous acetic acid. It forms dimers linked by hydrogen bonds in solution and gaseous commonly referred as phases. It is weakly acidic in nature.