Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, 99.8%, for analysis

Catalog No. p-4089397
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
20 L
Packaging:
Glass bottle
Plastic Can
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222142500 250 mL Glass bottle
AC222140010 1 L Glass bottle
AC222140025 2.5 L Glass bottle
AC222140050 5 L Plastic drum
AC222140200 20 L Plastic Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC222142500 Supplier Thermo Scientific Chemicals Supplier No. 222142500
Only null left
Add to Cart

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.0°C to 16.5°C
Boiling Point 117.0°C to 118.0°C
CAS Min % 99.75
CAS Max % 100.0
Assay Percent Range 99.8%
Linear Formula CH3COOH
Beilstein 02, 96
Fieser 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
Merck Index 15, 54
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide
Formula Weight 60.05
Percent Purity 99.8%
Grade Analytical
Density 1.0480g/mL
Quantity 250 mL
Flash Point 40°C
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.3700 to 1.3730
Residue after Evaporation 0.001% max.
Specific Gravity 1.048
Chemical Name or Material Acetic acid, For Analysis
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.