Learn More
Proteochem Inc Acetic Acid Ampules 10X1Ml (10/Pk)
Catalog No. NC0696191
Encompass_Preferred
Proteomics Grade Acetic Acid Ampules 10 x 1.0 mL (CAS Number: 64-19-7) recommended as an HPLC and LCMS mobile phase additive. Each Acetic Acid ampule contains 1.0 mL of Proteomics Grade Acetic Acid.
Catalog No. NC0696191 Supplier Proteochem Inc Supplier No. LC620010AMPS
Proteomics Grade Acetic Acid Ampules 10 x 1.0 mL (CAS Number: 64-19-7) recommended as an HPLC and LCMS mobile phase additive. Each Acetic Acid ampule contains 1.0 mL of Proteomics Grade Acetic Acid.