Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid concentrate, For 1L standard solution, 0.1 M CH3COOH (0.1N), Solstice

Catalog No. 6002659 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-026-59 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-026-59 Supplier Solstice Supplier No. 380501EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for 1L standard solution, 0.1 M CH3COOH (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Quantity 1 Pc.
Packaging Ampoule
Linear Formula CH3CO2H
UN Number NONH for all modes of transport
Beilstein 506007
Formula Weight 60.05g/mol
Chemical Name or Material Acetic acid concentrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.