Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid-D4, deuteration degree min. 99.5%, for NMR spectroscopy, MagniSolv™, MilliporeSigma™

Catalog No. M8150360010 Shop All MilliporeSigma Products
Change view
Click to view available options
Glass ampoule; 10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
M8150360010 Glass ampoule; 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M8150360010 Supplier MilliporeSigma Supplier No. 8.15036.0010

missing translation for 'validMainframeMsg'

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Ampule
Physical Form Liquid
Quantity 10 mL
Vapor Pressure 15.4 hPa (20°C)
Odor Stinging
Color Colorless
CAS 1186-52-3
Assay Percent Range 99.5% min.
Molecular Formula C2H4O2
MDL Number MFCD00051051
Synonym acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
InChI Key QTBSBXVTEAMEQO-GUEYOVJQSA-N
SMILES [2H]OC(=O)C([2H])([2H])[2H]
IUPAC Name (²H₃)ethan(²H)oic acid
Molecular Weight (g/mol) 64.08
PubChem CID 2723903
Formula Weight 64.08
Chemical Name or Material Acetic acid-D4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.