Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid (glacial), 100%, Suprapur™, MilliporeSigma™

Catalog No. M1000661000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1000661000 1 L
M1000660250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1000661000 Supplier MilliporeSigma Supplier No. 1.00066.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-19-7
Molecular Weight (g/mol) 60.05
Synonym Ethanoic acid

Specifications

pH 2.5 (50 g/L, H2O, 20°C)
Melting Point 17°C
Boiling Point 116 to 118°C (1013 hPa)
Density 1.05 g/cm3 (at 20°C)
Quantity 1 L
Explosion Limit 4 to 19.9% (V)
Flash Point 39°C
Ignition Point 485°C
Packaging Glass Bottle
Assay Percent Range ≥ 99.8%
Molecular Formula C2H4O2
Vapor Pressure 15.4 hPa (20°C)
Solubility Information 602.9 g/L Soluble
Show More Show Less
Recommended Storage Store at 15 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.